missing translation for 'onlineSavingsMsg'
Learn More

Complement Factor B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£292.00 - £433.00

Specifications

Antigen Complement Factor B
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
Research Discipline Immunology
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18632311
View Documents
Novus Biologicals
NBP2-76494-25ul
25 μL
£292.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18217139
View Documents
Novus Biologicals
NBP2-76494
100 μL
£433.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Complement Factor B Polyclonal specifically detects Complement Factor B in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Complement Factor B
Unconjugated
Immunology
PBS, pH 7.2, containing 40% glycerol with 0.02% sodium azide
629
This antibody was developed against Recombinant Protein corresponding to amino acids: VDDKEHSIKVSVGGEKRDLEIEVVLFHPNYNINGKKEAGIPEFYDYDVALIKLKNKLKYGQTIRPICLPCTEGTTRALRLPPTTTCQQQKEELLPAQDIKALFVSEEEKKLTRKEVYIKNGDKKGSCERDAQYAPGYDKVKDISEVV
Primary
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Polyclonal
Rabbit
Human
BF AHUS4, B-factor, properdin, BFD, C3 proaccelerator, C3 proactivator, C3/C5 convertase, complement factor B, EC 3.4.21, EC 3.4.21.47, FB, FBI12, GBGCFAB, Glycine-rich beta glycoprotein, glycine-rich beta-glycoprotein, H2-Bf, PBF2, Properdin factor B
CFB
IgG
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.