missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Complement Factor B Antibody (Genentech patent anti-Factor B) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28833-1mg
This item is not returnable.
View return policy
Description
Complement Factor B Monoclonal antibody specifically detects Complement Factor B in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Complement Factor B | |
| Monoclonal | |
| Unconjugated | |
| P00751 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| Genentech patent anti-Factor B | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| BF AHUS4, B-factor, properdin, BFD, C3 proaccelerator, C3 proactivator, C3/C5 convertase, complement factor B, EC 3.4.21, EC 3.4.21.47, FB, FBI12, GBGCFAB, Glycine-rich beta glycoprotein, glycine-rich beta-glycoprotein, H2-Bf, PBF2, Properdin factor B | |
| Complement Factor B | |
| 1 mg | |
| Immunology | |
| 629 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction