missing translation for 'onlineSavingsMsg'
Learn More

Complement Component C5aR1 Rabbit anti-Human, Clone: 32-G1, Novus Biologicals™

Product Code. 18384992 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
0.2 mg
Unit Size:
0.2mg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18384992 0.025 mg 25µg
18342483 0.2 mg 0.2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18384992 Supplier Novus Biologicals Supplier No. NBP3120850.025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Complement Component C5aR1 Monoclonal antibody specifically detects Complement Component C5aR1 in Human samples. It is validated for Western Blot, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Complement Component C5aR1
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone 32-G1
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation PBS
Gene Alias C5A, C5a anaphylatoxin chemotactic receptor, C5aR, C5a-R, C5ARC5a anaphylatoxin receptor, C5R1C5a ligand, CD88, CD88 antigen, complement component 5 receptor 1, complement component 5 receptor 1 (C5a ligand), complement component 5a receptor 1
Host Species Rabbit
Immunogen This antibody was raised by immunizing mice a synthetic Complement Component C5aR1 C-terminal phosphopeptide that contained phosphoserine instead of serine at position 334 and also at the neighboring phosphorylation sites Ser-332 and Ser-338.
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline GPCR, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 728
Target Species Human
Content And Storage Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C.
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.