missing translation for 'onlineSavingsMsg'
Learn More

Complement Component C5aR1 Antibody (20/70), Novus Biologicals™

Product Code. 18381312 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18381312 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18381312 Supplier Novus Biologicals Supplier No. NBP311236

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

Complement Component C5aR1 Monoclonal specifically detects Complement Component C5aR1 in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Functional.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Complement Component C5aR1
Applications Flow Cytometry, Immunofluorescence, Functional Assay
Classification Monoclonal
Clone 20/70
Concentration 0.1 mg/ml
Conjugate Unconjugated
Dilution Flow Cytometry, Immunocytochemistry/Immunofluorescence, Functional
Formulation 0.2 um filtered solution in PBS, 0.1% BSA
Gene Alias C5A, C5a anaphylatoxin chemotactic receptor, C5aR, C5a-R, C5ARC5a anaphylatoxin receptor, C5R1C5a ligand, CD88, CD88 antigen, complement component 5 receptor 1, complement component 5 receptor 1 (C5a ligand), complement component 5a receptor 1
Host Species Rat
Immunogen RBL-2H3 transfectants expressing murine C5aR on the cell surface
Purification Method Protein G purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline GPCR, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 728
Target Species Mouse
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.