missing translation for 'onlineSavingsMsg'
Learn More

Complement Component C5a Antibody (C17/5), Alexa Fluor™ 700, Novus Biologicals™

Product Code. 18417627 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18417627 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18417627 Supplier Novus Biologicals Supplier No. NBP223498AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Complement Component C5a Monoclonal antibody specifically detects Complement Component C5a in Human, Baboon samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Complement Component C5a
Applications Western Blot, ELISA
Classification Monoclonal
Clone C17/5
Conjugate Alexa Fluor 700
Formulation 50mM Sodium Borate
Gene Alias C5, C5A, HE
Host Species Mouse
Immunogen Purified human C5a
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Innate Immunity, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 727
Target Species Human, Baboon
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.