missing translation for 'onlineSavingsMsg'
Learn More

Complement Component C5a Antibody (295103) [DyLight 350], Novus Biologicals™

Product Code. 30648541 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30648541 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30648541 Supplier Novus Biologicals Supplier No. FAB2150D

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

Complement Component C5a Monoclonal antibody specifically detects Complement Component C5a in Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Complement Component C5a
Applications Western Blot
Classification Monoclonal
Clone 295103
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias C5, C5A, HE
Host Species Rat
Immunogen E. coli-derived recombinant mouse C5a, Asn679-Arg755, Accession # P06684
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Innate Immunity, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 727
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.