missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement Component C5a Antibody (215), mFluor Violet 450 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89578MFV450
This item is not returnable.
View return policy
Description
Complement Component C5a Monoclonal antibody specifically detects Complement Component C5a in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Complement Component C5a | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| 215 | |
| Immunohistochemistry-Paraffin | |
| C5, C5A, HE | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Complement Component C5a (Accession#: NP_001726.2; Leu679-Arg751). | |
| 0.1 mL | |
| Immunology, Innate Immunity, Stem Cell Markers | |
| 727 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction