missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Complement Component C5/C5a Monoclonal antibody specifically detects Complement Component C5/C5a in Human samples. It is validated for Sandwich ELISA Detection
Specifications
Specifications
| Antigen | Complement Component C5/C5a |
| Applications | Sandwich ELISA Detection |
| Classification | Monoclonal |
| Clone | 32 |
| Conjugate | Unconjugated |
| Dilution | Sandwich ELISA Detection 1:1000-1:10000 |
| Formulation | 0.2 um filtered solution in PBS |
| Gene Alias | C5a;C5b;C5D;Complement C5;CPAMD4;ECLZB |
| Host Species | Mouse |
| Immunogen | This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Complement Component C5/C5a (Accession#: NP_001726.2; Leu679-Arg751). |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?