missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement Component C3aR Antibody (hC3aRZ1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-64835
This item is not returnable.
View return policy
Description
Complement Component C3aR Monoclonal specifically detects Complement Component C3aR in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
| Complement Component C3aR | |
| Monoclonal | |
| 1 mg/mL | |
| Flow Cytometry 1:10-1:20, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| AZ3B, AZ3BC3a anaphylatoxin chemotactic receptor, C3a-R, C3AR, C3AR1, C3ARcomplement component 3 receptor 1, C3R1, complement component 3a receptor 1, Complement Component C3a R, Complement Component C3aR, HNFAG09 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 719 | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| hC3aRZ1 | |
| Unconjugated | |
| Q16581 | |
| C3AR1 | |
| Recombinant human C3aR protein | |
| 0.1 mg | |
| Primary | |
| NB100-64835 recognizes the receptor for human complement component C3a (C3aR), a 55kD cell surface protein of the rhodopsin family of G protein-coupled seven-transmembrane receptors. C3a is an anaphylatoxin, which is a potent proinflammatory mediator that possesses immunoregulatory properties. C3a regulates inflammatory functions by binding to the C3a receptor. C3aR is expressed on various tissues and leucocytes including basophils, neutrophils, mast cells, eosinophils and activated T lymphocytes. Expression levels of C3aR on human cells varies significantly between donors. |
|
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction