missing translation for 'onlineSavingsMsg'
Learn More

Complement Component C3aR Antibody (3G7), Alexa Fluor™ 700, Novus Biologicals™

Product Code. 18398180 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18398180 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18398180 Supplier Novus Biologicals Supplier No. NBP281062AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Complement Component C3aR Monoclonal specifically detects Complement Component C3aR in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Complement Component C3aR
Applications Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 3G7
Conjugate Alexa Fluor 700
Dilution Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias AZ3B, AZ3BC3a anaphylatoxin chemotactic receptor, C3a-R, C3AR, C3AR1, C3ARcomplement component 3 receptor 1, C3R1, complement component 3a receptor 1, Complement Component C3a R, Complement Component C3aR, HNFAG09
Gene Symbols C3AR1
Host Species Mouse
Immunogen The 2e loop of the Complement Component C3aR.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 719
Test Specificity Binds specifically to the 2e loop of the human C3a receptor, residues 185-193.
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.