missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Complement Component C2 Antibody (ARGX-117) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28215-1mg
This item is not returnable.
View return policy
Description
Complement Component C2 Monoclonal antibody specifically detects Complement Component C2 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Complement Component C2 | |
| Monoclonal | |
| Unconjugated | |
| P06681 | |
| Human | |
| Protein A purified | |
| RUO | |
| 717 | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Functional Assay | |
| ARGX-117 | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| C3/C5 convertase, CO2, complement C2, complement component 2, complement component C2, DKFZp779M0311, EC 3.4.21, EC 3.4.21.43 | |
| Complement C2 | |
| 1 mg | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction