missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement Component C2 Antibody (002), mFluor Violet 610 SE, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Complement Component C2 Monoclonal antibody specifically detects Complement Component C2 in Human samples. It is validated for ELISA
Specifications
Specifications
| Antigen | Complement Component C2 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 002 |
| Conjugate | mFluor Violet 610 SE |
| Dilution | ELISA |
| Formulation | 50mM Sodium Borate |
| Gene Alias | C3/C5 convertase, CO2, complement C2, complement component 2, complement component C2, DKFZp779M0311, EC 3.4.21, EC 3.4.21.43 |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human Complement Component C2 (Met 1-Leu 752; Accession#: NP_000054.2). |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?