missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement Component C1s Antibody (9), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-05023H
This item is not returnable.
View return policy
Description
Complement Component C1s Monoclonal specifically detects Complement Component C1s in Human samples. It is validated for ELISA, Immunohistochemistry.
Specifications
| Complement Component C1s | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| C1S | |
| C1s proenzyme purified from human plasma | |
| 0.1 mL | |
| Primary | |
| Specific for C1s proenzyme and activated free C1s | |
| Store at 4C in the dark. | |
| IgG1 κ |
| ELISA, Immunohistochemistry | |
| 9 | |
| ELISA, Immunohistochemistry | |
| basic proline-rich peptide IB-1, C1 esterase, complement C1s subcomponent, Complement component 1 subcomponent s, complement component 1, s subcomponent, EC 3.4.21, EC 3.4.21.42, FLJ44757 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 716 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction