missing translation for 'onlineSavingsMsg'
Learn More

Complement Component C1ra Antibody [FITC], Novus Biologicals™

Product Code. 30733580 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30733580 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30733580 Supplier Novus Biologicals Supplier No. AF7160F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Complement Component C1ra Polyclonal antibody specifically detects Complement Component C1ra in Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Complement Component C1ra
Applications Western Blot
Classification Polyclonal
Conjugate FITC
Formulation PBS
Gene Alias C1r;Complement C1r-A subcomponent;mC1;mC1rA;Peptidase S1 domain-containing protein
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Complement Component C1ra, Ser17-Asn707, Accession # AAH04637
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50909
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.