missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement Component C1r Antibody (OTI1F1), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-70318CL1
This item is not returnable.
View return policy
Description
Complement Component C1r Monoclonal antibody specifically detects Complement Component C1r in Canine,Human samples. It is validated for Western Blot
Specifications
| Complement Component C1r | |
| Monoclonal | |
| CoraFluor 1 | |
| complement C1r subcomponent, Complement component 1 subcomponent r, complement component 1, r subcomponent, EC 3.4.21, EC 3.4.21.41 | |
| Human recombinant protein fragment corresponding to amino acids 262-562 of human C1R (NP_001724) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Canine, Human | |
| Purified |
| Western Blot | |
| OTI1F1 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 715 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction