missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement Component C1qA Mouse anti-Human, Clone: C1QA/2955, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07733-100ug
This item is not returnable.
View return policy
Description
Complement Component C1qA Monoclonal specifically detects Complement Component C1qA in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin, Protein Array.
Specifications
| Complement Component C1qA | |
| Monoclonal | |
| 0.2 mg/mL | |
| Western Blot 1 to 2 μg/mL, Immunohistochemistry-Paraffin 1 to 2 μg/mL, Protein Array | |
| C1QA complement component 1, q subcomponent, A chain | |
| Recombinant fragment (around aa 104-237) of human Complement Component C1qA protein (exact sequence is proprietary) | |
| 100 μg | |
| Immunology, Innate Immunity | |
| 712 | |
| Store at 4°C. | |
| IgG2b κ |
| Western Blot, Immunohistochemistry (Paraffin), Peptide Array | |
| C1QA/2955 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction