missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement Component C1qA Mouse anti-Human, Clone: C1QA/2952, Novus Biologicals™
Mouse Monoclonal Antibody
£377.00
Specifications
| Antigen | Complement Component C1qA |
|---|---|
| Clone | C1QA/2952 |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin 1 to 2 μg/mL, Protein Array |
| Applications | Immunohistochemistry (Paraffin), Peptide Array |
Description
Complement Component C1qA Monoclonal specifically detects Complement Component C1qA in Human samples. It is validated for Immunohistochemistry-Paraffin, Protein Array.Specifications
| Complement Component C1qA | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin), Peptide Array | |
| Unconjugated | |
| Mouse | |
| Immunology, Innate Immunity | |
| 10 mM PBS with 0.05% BSA | |
| 712 | |
| IgG2b κ | |
| Protein A or G purified |
| C1QA/2952 | |
| Immunohistochemistry-Paraffin 1 to 2 μg/mL, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| C1QA complement component 1, q subcomponent, A chain | |
| Recombinant fragment (around aa 104-237) of human Complement Component C1qA protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title