Learn More
Invitrogen™ Complement C9 Monoclonal Antibody (22)

Mouse Monoclonal Antibody
Brand: Invitrogen™ ABS0042202
Description
ABS 004-22-02 is specific for complement factor C9. ABS 004-22-02 has been successfully used in ELISA and Western blot procedures. ABS 004-22-02 recognizes C9 in human serum diluted 1:50 in Tris buffer (20 mM Tris-base, 1 mM MgCl2, 1 mM CaCl2 and 140 mM NaCl) and incubated for 2 hours at 37°C using a human IgM coated (10 μg/mL overnight at 4°C, blocked with PBS 7.2 + 1% BSA for 1 hour) ELISA plate. NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL.
C5b-9 membrane attack complexes are assembled from five precursor molecules in the serum. Proteolytic cleavage of C5 by C5 convertase generates C5b which initiates assembly of the C5b-9 complex. The last step of C5b-9 complex formation involves polymerization of C9 which accompanies insertion of the complex into the cell membrane. During formation of C5b-8 and C9 polymerization, neoantigens are generated which are unique to the C5b-9 complex and are not present on any of the individual native complex components. The complement regulatory proteins CD59 and complement S-protein can both prevent C5b-9 insertion into the cell membrane. The formed SC5b-9 complex is unable to attach to cells and is cytolytically inactive. C5b-9 is involved in the progression of chronic proteinuric renal disease by mediating continuous tubulointerstitial damage. Early tubulointerstitial injury in the remnant kidney can be improved when C5b-9 complex forming is abrogated.
Specifications
| Complement C9 | |
| Monoclonal | |
| 1.03 mg/mL | |
| PBS with 15mM sodium azide; pH 7.4 | |
| P02748 | |
| C9 | |
| Complement component C9. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 22 | |
| Unconjugated | |
| C9 | |
| ARMD15; C9; C9D; complement C9; complement component 9; complement component C9; Complement component C9a; Complement component C9b; complement protein C9; complement protein C9 precursor | |
| Mouse | |
| Protein A | |
| RUO | |
| 735 | |
| 4°C, store in dark | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.