missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement C7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-00202-50ul
This item is not returnable.
View return policy
Description
Complement C7 Polyclonal antibody specifically detects Complement C7 in Human samples. It is validated for ELISA
Specifications
| Complement C7 | |
| Polyclonal | |
| ELISA 1:5000-1:10000 | |
| complement component 7, complement component C7 | |
| Produced in rabbits immunized with purified, recombinant Human Complement C7 (Accession#: P10643; Met1-Gln843) | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 730 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction