missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Complement C6 Monoclonal antibody specifically detects Complement C6 in Human samples. It is validated for ELISA
Specifications
Specifications
| Conjugate | mFluor Violet 610 SE |
| Dilution | ELISA |
| Formulation | 50mM Sodium Borate |
| Content And Storage | Store at 4C in the dark. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?