missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement C6 Antibody (037), DyLight 680, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90102FR
This item is not returnable.
View return policy
Description
Complement C6 Monoclonal specifically detects Complement C6 in Human samples. It is validated for ELISA.
Specifications
| Complement C6 | |
| Monoclonal | |
| DyLight 680 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 37 | |
| ELISA | |
| complement component 6, complement component C6 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Complement C6 (AAA59668.1; Met1-Ala934). | |
| 0.1 mL | |
| Immunology, Innate Immunity | |
| 729 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction