missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Complement C5 Antibody (lendalizumab) - Humanized, IgG2SA, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28576-50ug
This item is not returnable.
View return policy
Description
Complement C5 Monoclonal antibody specifically detects Complement C5 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Complement C5 | |
| Monoclonal | |
| Unconjugated | |
| P01031 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG2 |
| Flow Cytometry, ELISA, Functional Assay | |
| lendalizumab | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| anaphylatoxin C5a analog, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 4, C5, complement C5, complement component 5, CPAMD4MGC142298, FLJ17816, FLJ17822 | |
| Complement C5 | |
| 50 μg | |
| Immunology, Innate Immunity, Stem Cell Markers | |
| 727 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction