missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Complement C4d Monoclonal Antibody (LP69)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA518044
This item is not returnable.
View return policy
Description
.
C4 is a component of the classical complement pathway and is split by activated C1s to C4a and C4b. Together C2a and C4b forms the classical C3 convertase. The molecular mass of C4 is 204 kDa and it consists of 3 subunits: a (97 kDa), b (75 kDa) and g (33 kDa).
Specifications
| Complement C4d | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| P0C0L4, P0C0L5 | |
| C4A, C4B | |
| Complement C4d from normal human plasma. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
| LP69 | |
| Unconjugated | |
| C4A | |
| acidic C4; acidic complement C4; Basic complement C4; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 2; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 3; C4; c4 complement; C4A; C4a anaphylatoxin; C4A2; C4A3; C4A4; C4A6; C4AD; C4B; C4B_2; C4B1; C4B12; C4B2; C4B3; C4B5; C4b-A; C4b-B; C4BD; C4D; C4d-A; C4d-B; C4F; C4S; CH; Chido form of C4; CO4; Complement C4 beta chain; Complement C4 gamma chain; Complement C4-A; Complement C4-A alpha chain; Complement C4-B; Complement C4-B alpha chain; complement C4B1a; complement component 4A (Rodgers blood group); complement component 4B (Chido blood group); CPAMD2; CPAMD3; DADB-112B14.11; FLJ60561; MGC164979; MHC class III region complement; RG; Rodgers form of C4 | |
| Mouse | |
| Protein G | |
| RUO | |
| 100293534, 720, 721 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction