missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Complement C4b Monoclonal antibody specifically detects Complement C4b in Human samples. It is validated for Western Blot, ELISA, ELISA
Specifications
Specifications
| Antigen | Complement C4b |
| Applications | Western Blot, ELISA, Sandwich ELISA |
| Classification | Monoclonal |
| Clone | 2E3 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500, ELISA, Sandwich ELISA 1:100 to 1:2000 |
| Formulation | In 1x PBS, pH 7.4 |
| Gene Accession No. | NP_000583 |
| Gene Alias | basic C4, Basic complement C4, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 3, C4B1, C4B12, C4B2, C4B3, C4FMGC164979, CH, Chido form of C4, CO4C4B5, complement C4-B, complement C4B1a, complement component 4B, complement component 4B (Chido blood group), CPAMD3FLJ60561, EC 2.1.1.144, EC 2.7.11 |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?