missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement C3c Antibody (10-02A) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-64339
This item is not returnable.
View return policy
Description
Complement C3c Monoclonal specifically detects Complement C3c in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
| Complement C3c | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| AHUS5, ARMD9, ASP, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1, complement C3, complement component 3, complement component C3, CPAMD1acylation-stimulating protein cleavage product | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| NB100-64339 recognizes 135 kDa complement component 3c (C3c), present in blood serum. C3c is generated over the course of complement activation, where convertase C4b2a (classical pathway) and convertase C3bBb (alternative pathway) cleave C3 to C3b and C3a. C3b is further degraded into iC3b and C3dg/C3d. iC3b is then slowly cleaved (possibly by factor I) to form C3c (beta chain + alpha' chain fragment 1 + alpha' chain fragment 2), C3dg and C3f. C3 is crucial in the induction of tolerance generated when an antigen is introduced into immunoprivileged sites and this is exploited by pathogens and cancer cells to evade the immune system by inhibiting complement activation. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| 10-02A | |
| Unconjugated | |
| P01024 | |
| C3 | |
| Human C3 from normal serum, highly purified by chromatography. | |
| 0.05 mg | |
| Immunology, Innate Immunity, Neuronal Cell Markers, Neuroscience | |
| 718 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction