missing translation for 'onlineSavingsMsg'
Learn More

Complement C3a Polyclonal Antibody, Invitrogen™

Chicken Polyclonal Antibody

Brand:  Invitrogen PA19567

Code : LD

Additional Details : Weight : 0.01000kg

Product Code. 11548661

  • £430.00 / 100µg
Estimated Shipment: 11-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

PA1-9567 detects C3 alpha complement in mouse samples. This product may cross react with C3a from other mammalian species. PA1-9567 has been successfully used in Immunoblot and ELISA procedures. PA1-9567 binds to a strong band at about 9kDa (C3a), two weaker bands with one at about 115 kDa (C3 alpha) and the other at 25-36 kDa (unknown or partial C3 cleavage). PA1-9567 immunogen is mouse C3 complement protein aa 678-692, part of C3a. Store at 4oC for 1-2 weeks, and -20oC for long-term storage.

Complement component C3 plays a central role in the activation of complement system. Its activation is required for both classical and alternative complement activation pathways. People with C3 deficiency are susceptible to bacterial infection.
Specifications

Specifications

Complement C3a
Polyclonal
Unconjugated
C3
Acylation stimulating protein; acylation-stimulating protein cleavage product; AHUS5; AI255234; ARMD9; ASP; C3; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1; C3a; C3a anaphylatoxin; C3adesArg; C3b; C3bc; C3-beta-c; C3r1; Complement C3; complement C3 alpha chain; Complement C3 beta chain; complement C3 protein (GPC3) precursor; Complement C3b alpha' chain; Complement C3c alpha' chain fragment 1; Complement C3c alpha' chain fragment 2; Complement C3d fragment; Complement C3dg fragment; Complement C3f fragment; Complement C3g fragment; complement component 3; complement component C3 alpha-chain; complement component C3a; complement component C3b; CPAMD1; epididymis secretory sperm binding protein Li 62p; HEL-S-62p; HSE-MSF; Plp; prepro-C3
Chicken
Affinity chromatography
RUO
12266
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
ELISA, Western Blot
1 mg/mL
PBS with 0.02% sodium azide
P01027
C3
Mouse C3 complement protein aa 678-692, part of C3a.
100 μg
Primary
Mouse
Antibody
IgY
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers