Learn More
Complement C3a Monoclonal Antibody (2991), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA140157
Code : LD
Additional Details : Weight : 0.01000kg
Description
MA1-40157 detects C3a/C3a des-Arg from human samples. MA1-40157 has been successfully used in Western blot and ELISA applications. Positive control: Activated human serum or purified human C3a desArg protein; Negative control: For Western blot, any irrelevant protein can be used.
Complement component C3 plays a central role in the activation of complement system. Its activation is required for both classical and alternative complement activation pathways. People with C3 deficiency are susceptible to bacterial infection.Specifications
Complement C3a | |
Monoclonal | |
0.1 mg/mL | |
PBS with 0.1% BSA and 0.02% sodium azide | |
P01024 | |
C3 | |
Human C3a des-Arg | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Western Blot | |
2991 | |
Unconjugated | |
C3 | |
Acylation stimulating protein; acylation-stimulating protein cleavage product; AHUS5; AI255234; ARMD9; ASP; C3; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1; C3a; C3a anaphylatoxin; C3adesArg; C3b; C3bc; C3-beta-c; C3r1; Complement C3; complement C3 alpha chain; Complement C3 beta chain; complement C3 protein (GPC3) precursor; Complement C3b alpha' chain; Complement C3c alpha' chain fragment 1; Complement C3c alpha' chain fragment 2; Complement C3d fragment; Complement C3dg fragment; Complement C3f fragment; Complement C3g fragment; complement component 3; complement component C3 alpha-chain; complement component C3a; complement component C3b; CPAMD1; epididymis secretory sperm binding protein Li 62p; HEL-S-62p; HSE-MSF; Plp; prepro-C3 | |
Mouse | |
Protein G | |
RUO | |
718 | |
4° C | |
Liquid |