missing translation for 'onlineSavingsMsg'
Learn More

Complement C3a Antibody (D17/1), DyLight 594, Novus Biologicals™

Product Code. 18416597 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18416597 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18416597 Supplier Novus Biologicals Supplier No. NBP223497DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Complement C3a Monoclonal antibody specifically detects Complement C3a in Human samples. It is validated for Western Blot, ELISA, Sandwich ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Complement C3a
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone D17/1
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias Acylation Stimulating Protein, acylation-stimulating protein cleavage product, AHUS5, ARMD9, ASP, C3, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1, C3a, C3a anaphylatoxin, C3adesArg, C3b, C3bc, C3-beta-c, complement C3, Complement C3 alpha chain, Complement C3 beta chain, Complement C3b alpha' chain, Complement C3c alpha' chain fragment 1, Complement C3c alpha' chain fragment 2, Complement C3d fragment, Complement C3dg fragment, Complement C3f fragment, Complement C3g fragment, complement component 3, complement component C3, complement component C3a, complement component C3b, CPAMD1, EC 3.4.21.43, epididymis secretory sperm binding protein Li 62p, HEL-S-62p, prepro-C3
Host Species Mouse
Immunogen Complement C3a Antibody (D17/1) - BSA Free was developed against Human C3a.
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Apoptosis, Autoimmune Diseases, Diabetes Research, Immunology, Inflammation, Innate Immunity, Neuroscience, Signal Transduction, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 718
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.