missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Complement C3a Monoclonal specifically detects Complement C3a in Human samples. It is validated for ELISA, Western Blot.
Specifications
Specifications
| Antigen | Complement C3a |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 6E8 |
| Conjugate | APC |
| Dilution | Western Blot, ELISA |
| Formulation | PBS |
| Gene Alias | Acylation Stimulating Protein, acylation-stimulating protein cleavage product, AHUS5, ARMD9, ASP, C3, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1, C3a, C3a anaphylatoxin, C3adesArg, C3b, C3bc, C3-beta-c, complement C3, Complement C3 alpha chain, Complement C3 beta chain, Complement C3b alpha' chain, Complement C3c alpha' chain fragment 1, Complement C3c alpha' chain fragment 2, Complement C3d fragment, Complement C3dg fragment, Complement C3f fragment, Complement C3g fragment, complement component 3, complement component C3, complement component C3a, complement component C3b, CPAMD1, EC 3.4.21.43, epididymis secretory sperm binding protein Li 62p, HEL-S-62p, prepro-C3 |
| Host Species | Mouse |
| Immunogen | Recombinant netrin domain of human C3. UniProt: P01024 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?