Learn More
Complement C3 Antibody (11H9), mFluor Violet 610 SE, Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals NB200-540MFV610
Product Code. 18638449
  • £527.00 / 0.10mL
Description
Complement C3 Monoclonal antibody specifically detects Complement C3 in Mouse, E. coli samples. It is validated for Flow Cytometry, Flow (Intracellular)