missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement C3 Antibody (11H9), Janelia Fluor™ 646, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NB200-540JF646
This item is not returnable.
View return policy
Description
Complement C3 Monoclonal antibody specifically detects Complement C3 in Mouse, E. coli samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoassay, Flow (Intracellular), CyTOF
Specifications
| Complement C3 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| Acylation Stimulating Protein, acylation-stimulating protein cleavage product, AHUS5, ARMD9, ASP, C3, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1, C3a, C3a anaphylatoxin, C3adesArg, C3b, C3bc, C3-beta-c, complement C3, Complement C3 alpha chain, Complement C3 beta chain, Complement C3b alpha' chain, Complement C3c alpha' chain fragment 1, Complement C3c alpha' chain fragment 2, Complement C3d fragment, Complement C3dg fragment, Complement C3f fragment, Complement C3g fragment, complement component 3, complement component C3, complement component C3a, complement component C3b, CPAMD1, EC 3.4.21.43, epididymis secretory sperm binding protein Li 62p, HEL-S-62p, prepro-C3 | |
| This Complement C3 Antibody (11H9) was developed against C57BL/6 thymocytes saturated with rat anti-Thy-1 monoclonal antibody of IgG2b subclass (RmT1). | |
| 0.1 mL | |
| Angiogenesis, Apoptosis, Autoimmune Diseases, Immunology, Inflammation, Innate Immunity, Mesenchymal Stem Cell Markers, Neurodegeneration, Neuroscience, Signal Transduction | |
| 718 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 11H9 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Mouse, E. coli | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction