missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement C3 Antibody (11H9), Allophycocyanin, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NB200-540APC
This item is not returnable.
View return policy
Description
Complement C3 Monoclonal specifically detects Complement C3 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| Complement C3 | |
| Monoclonal | |
| APC | |
| PBS with 0.05% Sodium Azide | |
| C3 | |
| This Complement C3 Antibody (11H9) was developed against C57BL/6 thymocytes saturated with rat anti-Thy-1 monoclonal antibody of IgG2b subclass (RmT1). | |
| 0.1 mL | |
| Angiogenesis, Apoptosis, Autoimmune Diseases, Immunology, Inflammation, Innate Immunity, Mesenchymal Stem Cell Markers, Neurodegeneration, Neuroscience, Signal Transduction | |
| 718 | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| 11H9 | |
| Flow Cytometry | |
| AHUS5, ARMD9, ASP, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1, complement C3, complement component 3, complement component C3, CPAMD1acylation-stimulating protein cleavage product | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Mouse Complement C3 and its activation products, C3b, iC3b, C3d and C3dg | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction