missing translation for 'onlineSavingsMsg'
Learn More

Abcam Compensation Beads Kit (0.7-0.9 μm)

Product Code. 32114016 Shop All Abcam Products
Change view
Click to view available options
Quantity:
2 x 5 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32114016 2 x 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32114016 Supplier Abcam Supplier No. ab3226992x5mL
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Compensation Beads Kit (0.7-0.9 μm) (ab322699) is part of the sample prep & detection kits range. Abcam offers high-quality biological reagents and tools including antibodies, proteins, assays, cell lines and lysates.

Compensation Beads Kit (0.7-0.9 μm) (ab322699) is part of the sample prep & detection kits range. Abcam offers high-quality biological reagents and tools including antibodies, proteins, assays, cell lines and lysates.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Quantity 2 x 5 mL
Regulatory Status RUO
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.