Learn More
Palintest™ Comparator Reagents
Product Code. 11998926
Click to view available options
Analytes:
Alkalinity (CaCO3)
Chlorine DPD 2 (Monochloramine)
Copper
Nitrate
Phosphate (LR)
Zinc
Unit Size:
Each
Pack of 200
Pack of 250
Product Code. 11998926
Brand: Palintest™ AK148
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
  • Tablet form for maximum convenience and simplicity.
  • Colorimetric test.
Specifications
Analytes Zinc
For Use With (Equipment) Palintest comparator and disc
Packaging 250/Pk.
Quantity 1 Each
Test Method Zincon and an alkaline buffer methodology