Learn More
Hanna Instruments™ Compact Thermohygrometer
Brand: Hanna Instruments™ HI9565
Code : 04
Detalles adicionales : Weight : 0.10000kg
Especificaciones
Compact Thermohygrometer | |
0°C to 60°C | |
0.1°C | |
9V | |
with Dew Point and Calibration Data-Logging Probe |
0.1°C | |
±3°C | |
340 g | |
9 V |