Learn More
Descripción
- Compact gel imaging system for imaging DNA/RNA, stain free, visible protein gels and autorads.
- The InGenius 5 system is a powerful system with a groundbreaking 5M pixel camera that gives exquisite pixel resolution and unrivalled sensitivity in its class.
- The system uses a F1.4 manual lens to enable perfect gel imaging.
- The maximum viewing area is 20 x 24 cm, which is very large for such a small, compact unit.
- A flexible system with internal lighting options including a UV transilluminator (purchased separately) for working with DNA gels, stain free, and autorads.
- The overhead LED white lighting is included as standard for easy sample positioning and focusing.
- The InGenius 5 is a versatile system that has been designed for stain free applications. Its camera enables the sample to be exposed for extended periods.
- Safely excise your DNA bands from agarose gel with the darkroom door open using the optional safety shield protecting the user when working with UV.
- An intuitive system that permits images to be saved directly to USB or network for easy transfer.
- Quickly generate quantitative data with GeneTools analysis software.
- InGenius 5 comes with unlimited copies of GeneTools.
Especificaciones
Especificaciones
| Filter Area | 20 x 24 cm |
| For Use With (Application) | Gel Imaging |
| Product Type | Gel Imaging System |
| Display Type | None |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?