missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COMP/Thrombospondin-5 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£166.00 - £359.00
Specifications
| Antigen | COMP/Thrombospondin-5 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18620002
|
Novus Biologicals
NBP2-92758-0.02ml |
0.02 mL |
£166.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18615132
|
Novus Biologicals
NBP2-92758-0.1ml |
0.1 mL |
£359.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
COMP/Thrombospondin-5 Polyclonal antibody specifically detects COMP/Thrombospondin-5 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| COMP/Thrombospondin-5 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Signal Transduction, Stem Cells | |
| PBS with 50% glycerol, pH7.3. | |
| 1311 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| cartilage oligomeric matrix protein, cartilage oligomeric matrix protein (pseudoachondroplasia, epiphyseal dysplasia1, multiple), cartilage oligomeric matrix protein(pseudoachondroplasia, epiphyseal dysplasia1, multiple), EDM1, EPD1, MEDMGC131819, MGC149768, PSACH, pseudoachondroplasia (epiphyseal dysplasia 1, multiple), THBS5, Thrombospondin-5, TSP5 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 20-160 of human COMP (NP_000086.2). GQGQSPLGSDLGPQMLRELQETNAALQDVRELLRQQVREITFLKNTVMECDACGMQQSVRTGLPSVRPLLHCAPGFCFPGVACIQTESGARCGPCPAGFTGNGSHCTDVNECNAHPCFPRVRCINTSPGFRCEACPPGYSG | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title