missing translation for 'onlineSavingsMsg'
Learn More

COMP/Thrombospondin-5 Antibody (360215) [Janelia Fluor™ 549], Novus Biologicals™

Product Code. 30737294 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30737294 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30737294 Supplier Novus Biologicals Supplier No. FAB3134I

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

COMP/Thrombospondin-5 Monoclonal antibody specifically detects COMP/Thrombospondin-5 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen COMP/Thrombospondin-5
Applications Western Blot
Classification Monoclonal
Clone 360215
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias cartilage oligomeric matrix protein, cartilage oligomeric matrix protein (pseudoachondroplasia, epiphyseal dysplasia1, multiple), cartilage oligomeric matrix protein(pseudoachondroplasia, epiphyseal dysplasia1, multiple), EDM1, EPD1, MEDMGC131819, MGC149768, PSACH, pseudoachondroplasia (epiphyseal dysplasia 1, multiple), THBS5, Thrombospondin-5, TSP5
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human COMP/Thrombospondin-5, Gln21-Ala757 (Ala256Arg), Accession # P49747.2
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Signal Transduction, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 1311
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.