missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ COMP Monoclonal Antibody (C10), Invitrogen™

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA550899
This item is not returnable.
View return policy
Description
COMP is a noncollagenous extracellular matrix protein. It consists of five identical glycoprotein subunits, each with EGF like and calcium binding (thrombospondin like) domains. Oligomerization results from formation of a five stranded coiled coil and disulfides. Binding to other ECM proteins such as collagen appears to depend on divalent cations. Mutations can cause the osteochondrodysplasias pseudochondroplasia (PSACH) and multiple epiphyseal dysplasia (MED).
Specifications
| COMP | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| Q9R0G6 | |
| COMP | |
| Recombinant Mouse COMP protein, Gln554-Gln753 (Accession #Q9R0G6). | |
| 200 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2b κ |
| Western Blot, Immunohistochemistry (Paraffin), Western Blot | |
| C10 | |
| Unconjugated | |
| COMP | |
| Cartilage oligomeric matrix protein; cartilage oligomeric matrix protein (pseudoachondroplasia, epiphyseal dysplasia 1, multiple); cartilage oligomeric matrix protein(pseudoachondroplasia, epiphyseal dysplasia 1, multiple); Comp; EDM1; EPD1; MED; MGC131819; MGC149768; PSACH; pseudoachondroplasia (epiphyseal dysplasia 1, multiple); putative cartilage oligomeric matrix protein; THBS5; Thrombospondin5; Thrombospondin-5; TSP 5; TSP5 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 12845 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction