Learn More
Thermo Scientific™ Temperature Control Connectivity Accessories
Product Code. 10765689 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
1.8 m Interface Cable
5 ft. Interface Cable
5 m Interface Cable
Communication Extension Board
Software
For Use With (Equipment):
Cold/Heated Bath Circulators
RS-232 Communications Interface
Recirculating Chiller or Bath
Unit Size:
Each
Product Code. Type For Use With (Equipment) unitSize
10765689 Communication Extension Board Recirculating Chiller or Bath Each
11608048 Software RS-232 Communications Interface Each
11650991 1.8 m Interface Cable Cold/Heated Bath Circulators Each
11660991 5 ft. Interface Cable Cold/Heated Bath Circulators Each
10252278 5 m Interface Cable Cold/Heated Bath Circulators Each
5 options
Product Code. 10765689
Brand: Thermo Scientific™ 1600076
Only null left
Add to basket
Ajouter au panier
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Maximize the efficiency of your temperature control device with Thermo Scientific™ Temperature Control Connectivity and monitoring accessories.
Connect your Thermo Scientific temperature control solution with your application.
  • Available for temperature control devices equipped with communications
  • Designed to work with your Thermo Scientific recirculating chiller or bath equipped with communications
  • Available in popular configurations that match your device’s connections and interface with your process
  • Take the guess work out of finding parts to match your recirculating chiller or bath
Thermo Scientific™ NEScom™ Windows-based software
  • Provides remote control of Thermo Scientific units equipped with an RS-232 communications interface
  • Choose upper and lower temperature alarm limits and monitor system status with an alarm
  • Record your results on a user selectable graph
  • Select from easy–to–use product icons
  • Create graphs and charts
  • Easily configure ramping functions over time
  • View pop-up alarm windows which display the error condition
  • View a virtual controller screen
  • PC compatible only
TRUSTED_SUSTAINABILITY
Specifications
Communications Ethernet/LAN
For Use With (Equipment) Recirculating Chiller or Bath
Type Communication Extension Board