Learn More
Sartorius Stedim Biotech™ Communication Kit for Sterisart Universal Pumps
Product Code. 15932610 Shop All Sartorius Products
Click to view available options
Type:
Communication kit
Unit Size:
Each
Product Code. 15932610
Brand: Sartorius Stedim Biotech™ 1ZE0040
Only null left
Add to basket
Add to basket
Sartorius Stedim Biotech™ Communication Kit for Sterisart Universal Pumps contains all necessary cables and cards for device communication.
  • Includes cables and cards necessary for device communication.
    • RJ45 cable
    • Serial cable
    • MC card
    • MMC/SD card reader
TRUSTED_SUSTAINABILITY
Specifications
Includes RJ45 cable and serial cable for Sterisart™ Universal Pump, MC card and MMC/SD card reader for 16420
Type Communication kit