missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Common gamma Chain/IL-2 R gamma Rabbit anti-Mouse, Clone: 3000000000000, Novus Biologicals™
Rabbit Monoclonal Antibody
£200.00 - £464.00
Specifications
| Antigen | Common gamma Chain/IL-2 R gamma |
|---|---|
| Clone | 3000000000000 |
| Concentration | 1 mg/mL |
| Dilution | Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Block/Neutralize |
| Applications | Flow Cytometry, Immunofluorescence, Immunoprecipitation, Neutralization |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18328192
|
Novus Biologicals
NBP3-12038-0.025MG |
0.025 mg |
£200.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18035974
|
Novus Biologicals
NBP3-12038-0.2MG |
0.2 mg |
£464.00
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Common gamma Chain/IL-2 R gamma Monoclonal antibody specifically detects Common gamma Chain/IL-2 R gamma in Mouse samples. It is validated for Flow Cytometry, Immunofluorescence, Immunoprecipitation, NeutralizationSpecifications
| Common gamma Chain/IL-2 R gamma | |
| 1 mg/mL | |
| Flow Cytometry, Immunofluorescence, Immunoprecipitation, Neutralization | |
| Unconjugated | |
| RUO | |
| Mouse | |
| CD132, CD132 antigen, CIDX, combined immunodeficiency, X-linked, common cytokine receptor gamma chain, common gamma-chain, cytokine receptor common subunit gamma, gamma(c), IL-2 receptor subunit gamma, IL-2R subunit gamma, IL-2RG, IMD4, interleukin 2 receptor, gamma, Interleukin-2 receptor subunit gamma, p64, SCIDX, SCIDX1, severe combined immunodeficiency | |
| This antibody was raised by immunising Lewis rats with transfected rat myeloma cell line YB2/0, expressing murine cytoplasmic-tailless IL-2R common gamma-chain (YB2/0-myt). | |
| Primary | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
| 3000000000000 | |
| Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Block/Neutralize | |
| Monoclonal | |
| Rabbit | |
| Cancer | |
| PBS | |
| 3561 | |
| IgG κ | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title