missing translation for 'onlineSavingsMsg'
Learn More

Common gamma Chain/IL-2 R gamma Antibody (3E12), Alexa Fluor™ 488, Novus Biologicals™

Product Code. 18389763
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18389763 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18389763 Supplier Bio-Techne Supplier No. NBP312038AF488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Common gamma Chain/IL-2 R gamma Monoclonal antibody specifically detects Common gamma Chain/IL-2 R gamma in Mouse samples. It is validated for Flow Cytometry, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Common gamma Chain/IL-2 R gamma
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone 3000000000000
Conjugate Alexa Fluor 488
Dilution Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias CD132, CD132 antigen, CIDX, combined immunodeficiency, X-linked, common cytokine receptor gamma chain, common gamma-chain, cytokine receptor common subunit gamma, gamma(c), IL-2 receptor subunit gamma, IL-2R subunit gamma, IL-2RG, IMD4, interleukin 2 receptor, gamma, Interleukin-2 receptor subunit gamma, p64, SCIDX, SCIDX1, severe combined immunodeficiency
Host Species Rabbit
Immunogen This antibody was raised by immunising Lewis rats with transfected rat myeloma cell line YB2/0, expressing murine cytoplasmic-tailless IL-2R common gamma-chain (YB2/0-myt).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 3561
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.