missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Common beta Chain Antibody (CSL311), Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28274-100ug
This item is not returnable.
View return policy
Description
Common beta Chain Monoclonal antibody specifically detects Common beta Chain in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Common beta Chain | |
| Monoclonal | |
| Unconjugated | |
| P32927 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG4 |
| Flow Cytometry, ELISA, Functional Assay | |
| CSL311 | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| beta, low-affinity, CD131, CD131 antigen, CDw131, colony stimulating factor 2 receptor, beta, low-affinity(granulocyte-macrophage), cytokine receptor common subunit beta, GM-CSF/IL-3/IL-5 receptor common beta subunit, GM-CSF/IL-3/IL-5 receptor common beta-chain, IL3RB, interleukin 3 receptor/granulocyte-macrophage colony stimulating factor 3receptor, beta (high affinity) | |
| CSF2Rb/ CD131 | |
| 100 μg | |
| Phospho Specific | |
| 1439 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction