missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Common beta Chain Antibody (001), Alexa Fluor™ 405 , Novus Biologicals™
Shop All Bio Techne ProductsDescription
Common beta Chain Monoclonal antibody specifically detects Common beta Chain in Human samples. It is validated for ELISA
Specifications
Specifications
| Antigen | Common beta Chain |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 001 |
| Conjugate | Alexa Fluor 405 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | beta, low-affinity, CD131, CD131 antigen, CDw131, colony stimulating factor 2 receptor, beta, low-affinity(granulocyte-macrophage), cytokine receptor common subunit beta, GM-CSF/IL-3/IL-5 receptor common beta subunit, GM-CSF/IL-3/IL-5 receptor common beta-chain, IL3RB, interleukin 3 receptor/granulocyte-macrophage colony stimulating factor 3receptor, beta (high affinity) |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human Common beta Chain (NP_000386.1; Met 1-Trp 443). |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?