missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ COMMD8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557446
This item is not returnable.
View return policy
Description
Immunogen sequence: SRKDEIKQAL SREIVAISSA QLQDFDWQVK LALSSDKIAA LRMPLLSLHL DVKENGEVKP YSIEMSREEL QNLIQSLEAA NKVVLQLK Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 84%.
The protein encoded by this gene binds coiled-coil domain-containing protein 22 (CCDC22), and this complex can regulate the turnover of I-kappa-B and the activation of NF-kappa-B.
Specifications
| COMMD8 | |
| Polyclonal | |
| Unconjugated | |
| COMMD8 | |
| C77770; COMM domain containing 8; COMM domain-containing protein 8; Commd8; D5Buc26e; MDS022 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54951 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NX08 | |
| COMMD8 | |
| Recombinant protein corresponding to Human COMMD8. Recombinant protein control fragment (Product #RP-96728). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction