missing translation for 'onlineSavingsMsg'
Learn More

COMMD7, Mouse anti-Human, Clone: 1B9, Abnova™

Mouse Monoclonal Antibody

Brand:  Abnova H00149951-M01.100ug

Product Code. 16118573

  • £303.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Sequence: MGRLHCTEDPVPEAVGGDMQQLNQLGAQQFSALTEVLFHFLTEQKEVERFLAQLSEFATTNQISLGSLRSIVKSLLLVPNGALKKSLTAKQVQADFITLGLSEEKATYFSEKWKQNTPTLARWAIGQTLMINQLIDMEWKFGVTSGSSELEKVGSIFLQLKLVVKKGNQTKNVYIELTLPQFYSFLHEMERVRTSMECFC
Specifications

Specifications

COMMD7
Monoclonal
Unconjugated
COMM domain containing 7
C20orf92/FLJ14987/MGC33315/dJ1085F17.3
Mouse
100 μg
Primary
Human
Antibody
ELISA, Western Blot
1B9
In 1x PBS, pH 7.4
BC047440
COMMD7
COMMD7 (AAH47440, 1 a.a. ∼ 200 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
RUO
149951
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.