missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ COMMD7 Antibody Blocking Peptide

Product Code. 18665876 Shop All Bio Techne Products
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
This item is not returnable. View return policy

Product Code. 18665876

Brand: Novus Biologicals™ NBP242124PEP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Blocking Peptide

TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 149951
Species Human
Concentration 0.2 mg/mL
Content And Storage Store at -20°C. Avoid freeze/thaw cycles.
For Use With (Application) This Peptide is Useful as a Blocking Peptide for NBP2-42124. It Usually Blocks The Antibody Activity Completely by Incubating The Peptide with Equal Volume of Antibody for 30 Min (at 37°C.)
Gene Alias C20orf92, chromosome 20 open reading frame 92, COMM domain containing 7, COMM domain-containing protein 7, dJ1085F17.3, FLJ14987, MGC33315
Gene Symbol COMMD7
Quantity 0.05 mg
Research Category Cancer, Cell Cycle and Replication
Storage Buffer PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.