missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ COMMD5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583847
This item is not returnable.
View return policy
Description
Immunogen sequence: TFRDQLQELC IPQDLVGDLA SVVFGSQRPL LDSVAQQQGA WLPHVADF Highest antigen sequence indentity to the following orthologs: Mouse - 73%, Rat - 73%.
COMMD5 is a protein coding gene. May modulate activity of cullin-RING E3 ubiquitin ligase (CRL) complexes. Negatively regulates cell proliferation. Negatively regulates cell cycle G2/M phase transition probably by transactivating p21/CDKN1A through the p53/TP53-independent signaling pathway. Involved in kidney proximal tubule morphogenesis. Down-regulates activation of NF-kappa-B.
Specifications
| COMMD5 | |
| Polyclonal | |
| Unconjugated | |
| COMMD5 | |
| 2310065H03Rik; AI854466; COMM domain containing 5; COMM domain-containing protein 5; COMMD5; D15Ertd81e; FLJ13008; HCARG; HT002; hypertension-related calcium regulated; hypertension-related calcium-regulated gene protein; hypertension-related, calcium regulated | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 28991 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunofluorescence | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9GZQ3 | |
| COMMD5 | |
| Recombinant protein corresponding to Human COMMD5. Recombinant protein control fragment (Product #RP-99259). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction