missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ COMMD3 Monoclonal Antibody (2E2)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA521467
This item is not returnable.
View return policy
Description
Peptide Sequence: MELSESVQKG FQMLADPRSF DSNAFTLLLR AAFQSLLDAQ ADEAVLDHPD LKHIDPVVLK HCHAAAATYI LEAGKHRADK STLSTYLEDC KFDRERIELF CTEYQNNKNS LEILLGSIGR SLPHITDVSW RLEYQIKTNQ LHRMYRPAYL VTLSVQNTDS PSYPEISFSC SMEQLQDLVG KLKDASKSLE RATQS.
COMMD3 may modulate activity of cullin-RING E3 ubiquitin ligase (CRL) complexes. may down-regulate activation of NF-kappa-B. Modulates Na(+) transport in epithelial cells by regulation of apical cell surface expression of amiloride-sensitive sodium channel (ENaC) subunits.
Specifications
| COMMD3 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q9UBI1 | |
| COMMD3 | |
| COMMD3 (AAH22898, 1 a.a. ~ 195 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Immunocytochemistry | |
| 2E2 | |
| Unconjugated | |
| COMMD3 | |
| AW550818; Bmi1 upstream; bmi-1 upstream gene protein; Bup; Bup protein; C10orf8; COMM domain containing 3; COMM domain-containing protein 3; COMMD3; D2Ertd542e; Protein Bup; Protein PIL | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 23412 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction